Anti-TRIM69 Rabbit Polyclonal Antibody
Catalog # PRSI28-186
Supplier: ProSci Inc.
Specifications
- Antigen name:Tripartite Motif Containing 69
- Antigen symbol:TRIM69
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Immunogen:A synthetic peptide corresponding a region of human RNF36.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
RNF36 is specifically expressed in germ cells at round spermatid stages during spermatogenesis. The protein encoded by this gene contains an N-terminal RING finger motif, a B-box, and a C-terminal B-30.2-like domain. When overexpressed in COS-7 and HEK-293 cells, this protein was shown to induce cell apoptosis.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TRIM69
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human