Order Entry
Italy
ContactUsLinkComponent
Anti-NR2F6 Rabbit Polyclonal Antibody
Anti-NR2F6 Rabbit Polyclonal Antibody
  PRSI27-531
undefined
Anti-NR2F6 Rabbit Polyclonal Antibody
  PRSI27-531

 

  • Antigen name:
    Nuclear Receptor Subfamily 2, Group F, Member 6
  • Antigen symbol:
    NR2F6
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    EAR2|EAR-2|ERBAL2
  • Immunogen:
    A synthetic peptide corresponding a region of human NR2F6.
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

Orphan nuclear receptor EAR-2 (NR2F6, V-erbA related protein EAR-2 ) is predicted to be a protein similar in primary structure to receptors for steroid hormones or thyroid hormone (T3).

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NR2F6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human