Specifications
- Antigen name:General Transcription Factor 2I
- Antigen symbol:GTF2I
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Dog,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:SPIN|DIWS|BTKAP1|WBSCR6|IB291|WBS|TFII-I|BAP135|GTFII-I
- Immunogen:A synthetic peptide corresponding a region of human GTF2I.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
GTF2I encodes a multifunctional phosphoprotein with roles in transcription and signal transduction. It is deleted in Williams-Beuren syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at chromosome 7q11.23. The exon (s) encoding 5' UTR has not been fully defined, but this gene is known to contain at least 34 exons, and its alternative splicing generates 4 transcript variants.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: GTF2I
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Dog