Anti-MSX1 Rabbit Polyclonal Antibody
Catalog # PRSI27-333
Supplier: ProSci Inc.
Specifications
- Antigen name:Msh homeobox 1
- Antigen symbol:MSX1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Dog
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:msh homeobox homolog 1|HYD1|HOX7|homeobox protein Hox-7|homeobox protein MSX-1|homeobox 7|msh homeobox 1-like protein|msh homeo box 1|STHAG1
- Immunogen:A synthetic peptide corresponding a region of human MSX1.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Slightly proximal to the Huntington disease locus, the human MSX1 gene is deleted in patients with Wolf-Hirschhorn syndrome. This gene is also called HOX7. The Msx family of vertebrate HOX genes was originally isolated by homology to the Drosophila msh (muscle segment homeo box) gene. This is a candidate gene for human cleft palate.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: MSX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human