Order Entry
Italy
ContactUsLinkComponent
Anti-FAM219A Rabbit Polyclonal Antibody
Anti-FAM219A Rabbit Polyclonal Antibody
Catalog # PRSI27-034
Supplier:  ProSci Inc.
undefined
Anti-FAM219A Rabbit Polyclonal Antibody
Catalog # PRSI27-034
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Family With Sequence Similarity 219, Member A
  • Antigen symbol:
    FAM219A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    bA573M23.5|C9orf25
  • Immunogen:
    A synthetic peptide corresponding a region of human C9orf25.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

The function of this protein has not been determined.The protein encoded by this gene has homologs that have been identified in mouse and macaque. The mouse and human proteins have a putative prenyl group binding site (CAAX box) at their C-terminus. A diverse list of proteins are known or strongly presumed to be the target of post-translational modification by the attachment of either a farnesyl or a geranyl-geranyl group to a cysteine residue at the C-terminus. The function of this protein has not been determined.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FAM219A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat