Anti-C7orf61 Rabbit Polyclonal Antibody
Catalog # PRSI26-861
Supplier: ProSci Inc.
Specifications
- Antigen symbol:C7orf61
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Immunogen:A synthetic peptide corresponding a region of human LOC402573.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
The exact function of LOC402573 remains unknown.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: 0
Antigen: C7orf61
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human