- Antigen name:Acyl-CoA Synthetase Bubblegum Family member 2
- Antigen symbol:ACSBG2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Immunogen:A synthetic peptide corresponding a region of human ACSBG2.
- Size:50 µg
- Pk:50 µG
ACSBG2 belongs to the ATP-dependent AMP-binding enzyme family, bubblegum subfamily.ACSBG2 mediates activation of long-chain fatty acids for both synthesis of cellular lipids, and degradation via beta-oxidation. It is able to activate long-chain fatty acids. Also able to activate very long-chain fatty acids; however, the relevance of such activity is unclear in vivo. ACSBG2 has increased ability to activate oleic and linoleic acid. It may play a role in spermatogenesis.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ACSBG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human