Anti-ARL13B Rabbit Polyclonal Antibody
PRSI26-696
- Antigen name:ADP-Ribosylation Factor-Like 13B
- Antigen symbol:ARL13B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:ARL2L1|JBTS8
- Immunogen:A synthetic peptide corresponding a region of human ARL13B.
- Size:50 µg
- Pk:50 µG
ARL13B has an evolutionarily conserved role mediating cilia function in multiple organs. N and C domains of ARL13B cooperatively regulate its ciliary localization and that N domain-dependent self-association of Arl13b may be important for its function in cilia biogenesis.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ARL13B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human