- Antigen name:Asparagine-Linked Glycosylation 1
- Antigen symbol:ALG1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:GDP-Mannose-Dolichol Diphosphochitobiose Mannosyltransferase|hMat-1|HMT1|CDG1K|Chitobiosyldiphosphodolichol Beta-Mannosyltransferase|HMT-1|HMAT1|Mat-1|MT-1
- Immunogen:A synthetic peptide corresponding a region of human ALG1.
- Size:50 µg
- Pk:50 µG
ALG1 catalyzes the first mannosylation step in the biosynthesis of lipid-linked oligosaccharides. Defects in ALG1 are the cause of congenital disorder of glycosylation type 1K (CDG1K).The biosynthesis of lipid-linked oligosaccharides is highly conserved among eukaryotes and is catalyzed by 14 glycosyltransferases in an ordered stepwise manner. Mannosyltransferase I (MT I) catalyzes the first mannosylation step in this process.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ALG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human