Order Entry
Italy
ContactUsLinkComponent
Anti-ST8 Alpha-N-Acetyl-Neuraminide Alpha-2,8-Sialyltransferase 4 Rabbit Polyclonal Antibody
Anti-ST8 Alpha-N-Acetyl-Neuraminide Alpha-2,8-Sialyltransferase 4 Rabbit Polyclonal Antibody
Catalog # PRSI26-335
Supplier:  ProSci Inc.
undefined
Anti-ST8 Alpha-N-Acetyl-Neuraminide Alpha-2,8-Sialyltransferase 4 Rabbit Polyclonal Antibody
Catalog # PRSI26-335
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    St8 Alpha-N-Acetyl-Neuraminide Alpha-2,8-Sialyltransferase 4
  • Antigen symbol:
    ST8SIA-4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    PST|ST8SIA-IV|SIAT8D|PST1
  • Immunogen:
    A synthetic peptide corresponding a region of human ST8SIA4.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

ST8SIA4 catalyzes the polycondensation of alpha-2,8-linked sialic acid required for the synthesis of polysialic acid, a modulator of the adhesive properties of neural cell adhesion molecule (NCAM1). ST8SIA4, a member of glycosyltransferase family 29, is a type II membrane protein that may be present in the Golgi apparatus.The protein encoded by this gene catalyzes the polycondensation of alpha-2,8-linked sialic acid required for the synthesis of polysialic acid, a modulator of the adhesive properties of neural cell adhesion molecule (NCAM1). The encoded protein, which is a member of glycosyltransferase family 29, is a type II membrane protein that may be present in the Golgi apparatus. Two transcript variants encoding different isoforms have been found for this gene.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ST8 Alpha-N-Acetyl-Neuraminide Alpha-2,8-Sialyltransferase 4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat