Anti-MARCH7 Rabbit Polyclonal Antibody
PRSI25-847
- Antigen name:Membrane-associated ring Finger (C3HC4) 7, E3 ubiquitin protein ligase
- Antigen symbol:MARCH7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:AXOT|RNF177|AXO|MARCH-VII
- Immunogen:A synthetic peptide corresponding a region of human MARCH7.
- Size:50 µg
- Pk:50 µG
MARCH7 is an E3 ubiquitin-protein ligase which may specifically enhance the E2 activity of HIP2. E3 ubiquitin ligases accept ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfer the ubiquitin to targeted substrates.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: MARCH7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat