Order Entry
Italy
ContactUsLinkComponent
Anti-MPP5 Rabbit Polyclonal Antibody
Anti-MPP5 Rabbit Polyclonal Antibody
Catalog # PRSI25-802
Supplier:  ProSci Inc.
undefined
Anti-MPP5 Rabbit Polyclonal Antibody
Catalog # PRSI25-802
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Membrane Palmitoylated Protein 5
  • Antigen symbol:
    MPP5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    MAGUK p55 subfamily member 5|PALS 1|Protein associated with Lin-7|Stardust Drosophila|MPP 5|Stardust.|FLJ12615|MPP5|MPP5_HUMAN|PALS1|Membrane protein palmitoylated 5
  • Immunogen:
    A synthetic peptide corresponding a region of human MPP5.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Members of the peripheral membrane-associated guanylate kinase (MAGUK) family function in tumor suppression and receptor clustering by forming multiprotein complexes containing distinct sets of transmembrane, cytoskeletal, and cytoplasmic signaling proteins. All MAGUKs contain a PDZ-SH3-GUK core and are divided into 4 subfamilies, DLG-like, ZO1-like, p55-like, and LIN2-like, based on their size and the presence of additional domains. MPP5 is a member of the p55-like MAGUK subfamily.Members of the peripheral membrane-associated guanylate kinase (MAGUK) family function in tumor suppression and receptor clustering by forming multiprotein complexes containing distinct sets of transmembrane, cytoskeletal, and cytoplasmic signaling proteins. All MAGUKs contain a PDZ-SH3-GUK core and are divided into 4 subfamilies, DLG-like (see DLG1; MIM 601014), ZO1-like (see TJP1; MIM 601009), p55-like (see MPP1; MIM 305360), and LIN2-like (see CASK; MIM 300172), based on their size and the presence of additional domains (Tseng et al., 2001 [PubMed 11311936]). MPP5 is a member of the p55-like MAGUK subfamily.[supplied by OMIM]. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MPP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat