Anti-EIF4G3 Rabbit Polyclonal Antibody
Catalog # PRSI25-620
Supplier: ProSci Inc.
Specifications
- Antigen symbol:EIF4G3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:eIF4GII|eIF4G 3|eIF-4G 3
- Immunogen:A synthetic peptide corresponding a region of human EIF4G3.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
EIF4G3 is probable component of the protein complex eIF4F, which is involved in the recognition of the mRNA cap, ATP-dependent unwinding of 5'-terminal secondary structure and recruitment of mRNA to the ribosome. It is thought to be a functional homolog of EIF4G1.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: EIF4G3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat