Order Entry
Italy
ContactUsLinkComponent
Anti-SLC26A3 Rabbit Polyclonal Antibody
Anti-SLC26A3 Rabbit Polyclonal Antibody
Catalog # PRSI25-021
Supplier:  ProSci Inc.
undefined
Anti-SLC26A3 Rabbit Polyclonal Antibody
Catalog # PRSI25-021
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Solute Carrier Family 26 Member A3
  • Antigen symbol:
    SLC26A3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Immunogen:
    A synthetic peptide corresponding a region of human SLC26A3.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

SLC26A3 is a transmembrane glycoprotein that transports chloride ions across the cell membrane in exchange for bicarbonate ions. It is localized to the mucosa of the lower intestinal tract, particularly to the apical membrane of columnar epithelium and some goblet cells. The protein is essential for intestinal chloride absorption, and mutations in this gene have been associated with congenital chloride diarrhea.The protein encoded by this gene is a transmembrane glycoprotein that functions as a sulfate transporter. It is localized to the mucosa of the lower intestinal tract, particularly to the apical membrane of columnar epithelium and some goblet cells. Mutations in this gene have been associated with congenital chloride diarrhea. Please see the Entrez Gene record to access additional publications.

SLC26A3 antibody can be used for detection of SLC26A3 by ELISA at 1:312500. SLC26A3 antibody can be used for detection of SLC26A3 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SLC26A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human