Order Entry
Italy
ContactUsLinkComponent
Anti-SIGLEC15 Rabbit Polyclonal Antibody
Anti-SIGLEC15 Rabbit Polyclonal Antibody
Catalog # PRSI6765
Supplier:  ProSci Inc.
undefined
Anti-SIGLEC15 Rabbit Polyclonal Antibody
Catalog # PRSI6765
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Sialic Acid Binding Ig-like Lectin 15
  • Antigen symbol:
    SIGLEC15
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Antigen synonyms:
    Siglec-15|SIGLEC-15|CD33L3|CD33 antigen-like 3|SIGLEC15|Sialic acid-binding Ig-like lectin 15|HsT1361
  • Storage buffer:
    PBS containing 0.02% sodium azide
  • Immunogen:
    SIGLEC15 antibody was raised against a 15 amino acid synthetic peptide near the amino terminus of human SIGLEC15.
  • Tested applications:
    E, WB, IHC, IF
  • Purification:
    Affinity chromatography purified via peptide column
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

SIGLEC15 Antibody: Siglecs are vertebrate cell-surface lectins that recognize sialylated glycans and are involved in many physiological processes, such as glycoprotein turnover, cellular trafficking, and pathogen recognition. Most Siglecs are expressed on cells of the immune system. SIGLEC15 is a type-I transmembrane protein consisting of two immunoglobulin (Ig)-like domains, a transmembrane domain containing a lysine residue, and a short cytoplasmic tail. SIGLEC15 can interact with the activating adaptor molecules DAP12/10. Its activating signaling potential and unique preference for glycan recognition implies that SIGLEC15 may be involved in the immune surveillance of tumors and probably plays a conserved, regulatory role in the immune system of vertebrates.

Type:
Antigen: SIGLEC15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat