Specifications
- Antigen name:Musashi RNA-binding Protein 2
- Antigen symbol:MSI2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Antigen synonyms:RNA-binding protein Musashi homolog 2|MSI2H|musashi-2
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:MSI2 antibody was raised against a 15 amino acid synthetic peptide near the amino terminus of human MSI2.
- Tested applications:E, WB, ICC, IF
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
Specifications
About this item
MSI2 Antibody: Musashi2 (MSI2) is an RNA-binding protein that is highly expressed in precursor cells in the ventricular and subventricular zones of the developing mammalian CNS. Like the related MSI1, MSI2 has been suggested to be involved stem cell production and maintenance. MSI2 is the predominant MSI protein in hematopoietic stem cells, and its knockdown leads to reduced engraftment and depletion in vivo. Expression levels of MSI2 are elevated in myeloid leukemia cells lines, and MSI2 appears to cooperate with BCR-ABL1 to induce an aggressive leukemia; the level of MSI2 directly correlates with decreased survival in patients. MSI2 negatively regulates the asymmetric cell fate determinant NUMB, suggesting that this signaling pathway may provide future targets for future therapies.
Type:
Antigen: MSI2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat