Order Entry
Italy
ContactUsLinkComponent
Anti-ATP2C1 Rabbit Polyclonal Antibody
Anti-ATP2C1 Rabbit Polyclonal Antibody
Catalog # PRSI6287
Supplier:  ProSci Inc.
undefined
Anti-ATP2C1 Rabbit Polyclonal Antibody
Catalog # PRSI6287
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    ATPase, Ca++ transporting, type 2C, member 1
  • Antigen symbol:
    ATP2C1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Antigen synonyms:
    ATP2C1
  • Storage buffer:
    PBS containing 0.02% sodium azide
  • Immunogen:
    ATP2C1 antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human ATP2C1.
  • Tested applications:
    E, WB, IHC, IF
  • Purification:
    Affinity chromatography purified via peptide column
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

ATP2C1 Antibody: ATP2C1, also known as secretory pathway Ca2+/Mn2+-ATPase (SPCA) 1, belongs to the family of P-type cation transport ATPases. This magnesium-dependent enzyme catalyzes the hydrolysis of ATP coupled with the transport of the calcium from the cytosol to the Golgi lumen. Defects in this gene cause Hailey-Hailey disease, an autosomal dominant disorder characterized by persistent blisters and erosions of the skin. Unlike the related protein ATP2C2, ATP2C1 is ubiquitously expressed and displays a lower maximal turnover rate for overall Ca2+-ATPase reaction and a higher apparent affinity for cytosolic Ca2+ activation of phosphorylation. Recent evidence suggests that ATP2C1 is a key regulator of insulin-like growth factor receptor (IGF1R) processing in tumor progression in basal breast cancers.

Type:
Antigen: ATP2C1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse