Anti-PPAPDC3 Rabbit Polyclonal Antibody
Product Details & Documents
PPAPDC3 Antibody: PPAPDC3, also known as nuclear envelope transmembrane protein 39 (NET39), was initially discovered in an in silico screen for secreted or membrane proteins. It is a member of the PAP2 superfamily of phosphatases and haloperoxidases. PPAPDC3 has recently been shown to act as a negative regulator of myoblast differentiation by diminishing the activity of the mammalian target of rapamycin TOR. PPAPDC3 is highly expressed in cardiac and skeletal muscle and becomes strongly upregulated during cultured myoblast differentiation tissues. Overexpression of PPAPDC3 in myoblasts repressed myogenesis while knockdown by RNA interference promoted differentiation indicating its part in the regulatory mechanism for myogenesis.
Type:
Antigen: PPAPDC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Documents
89417-256.pdf
Datasheet
Specifications
- Size:100 µg
- Western blot:Yes
- Clonality:Polyclonal
- Purification:Affinity chromatography purified via peptide column
- ImmunoFluorescence:Yes
- Isotype:IgG
- Host:Rabbit
- Antigen name:Phosphatidic Acid Phosphatase Type 2 Domain Containing 3
- Cat. No.:PRSI5519
- Reactivity:Human|Rat|Mouse
- Antigen synonyms:C9orf67|NET39|Phosphatidic acid phosphatase type 2 domain-containing protein 3|Probable lipid phosphate phosphatase PPAPDC3|PPAPDC3
- Immunogen:PPAPDC3 antibody was raised against a 19 amino acid synthetic peptide from near the amino of terminus human PPAPDC3.
- Conjugation:Unconjugated
- Tested applications:E, WB, IF
- Antigen symbol:PPAPDC3
- Storage buffer:PBS containing 0.02% sodium azide
- ELISA:Yes



