Order Entry
Italy
ContactUsLinkComponent
Anti-LGP2 Rabbit Polyclonal Antibody
Anti-LGP2 Rabbit Polyclonal Antibody
Catalog # PRSI4353
Supplier:  ProSci Inc.
undefined
Anti-LGP2 Rabbit Polyclonal Antibody
Catalog # PRSI4353
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    DEXH (Asp-Glu-X-His) box Polypeptide 58
  • Antigen symbol:
    LGP2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Antigen synonyms:
    DEXH (Asp-Glu-X-His) box polypeptide 58 |RNA helicase LGP2 |D11LGP2 |DHX58|D11lgp2e |LGP2|ortholog of mouse D11lgp2
  • Storage buffer:
    PBS containing 0.02% sodium azide
  • Immunogen:
    LGP2 antibody was raised against a 14 amino acid synthetic peptide from near the amino terminus of human LGP2.
  • Tested applications:
    E, WB
  • Purification:
    Affinity chromatography purified via peptide column
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

LGP2 Antibody: Anti-viral innate immune responses are triggered by pathogen-associated molecular patters (such as the accumulation of intracellular nucleic acids resulting from virus infections) and represent the first line of defense against numerous infectious organisms. The Toll-like receptors (TLR) 3, 7, 8, and 9 are expressed in immune cells and function as transmembrane pattern recognition receptors to detect foreign nucleic acids. Other proteins that play similar roles, such as RIG-1 and MDA5 are members of a CARD-helicase family and are expressed in the cytoplasm. A third protein, LGP2, is similar to RIG-1 and MDA5, except for lacking a homologous CARD domain. It is thought to act as an element of negative-feedback regulation of intracellular antiviral signaling. At least four isoforms of LGP2 are known to exist.

Type:
Antigen: LGP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat