Specifications
- Antigen name:BR serine/threonine kinase 2
- Antigen symbol:BRSK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Antigen synonyms:SADA|SAD1|PEN11B|Brain-selective kinase 2|C11orf7|STK29|HUSSY-12
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:BRSK2 antibody was raised against a 23 amino acid synthetic peptide from near the carboxy terminus of human BRSK2.
- Tested applications:E, WB, IHC, IF
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
Specifications
About this item
BRSK2 Antibody: BRSK2 was initially identified through a computer screen of the human genome and shows significant homology to the C. elegans neuronal cell polarity regulator SAD1. BRSK2 is expressed in the brain and to a lesser extent in the testes. BRSK2 is a member of the AMP-activated protein kinase subfamily and can be activated by the tumor suppressor kinase LKB1. More recently, it has been shown that both BRSK2 and the related protein BRSK1 are required for mammalian neuronal polarization. While BRSK1- and BRSK2-null mice were viable, double-mutant mice died within two hours of birth. Neurons from these mice showed uniformly-sized neurites as opposed to the normal long axon and multiple shorter dendrites. These neurites also displayed both axonal and dendritic markers. BRSK2 has also been shown to be an autoantigen in paraneoplastic limbic encephalitis. At least four isoforms of BRSK2 are known to exist.
Type:
Antigen: BRSK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse