Order Entry
Italy
Orders LinkContactUsLinkComponent
Anti-IPAF Rabbit Polyclonal Antibody

Anti-IPAF Rabbit Polyclonal Antibody

100 µg
Catalog # PRSI3107
Supplier:   ProSci Inc.
Anti-IPAF Rabbit Polyclonal Antibody
Catalog # PRSI3107
Supplier:  ProSci Inc.
Anti-IPAF Rabbit Polyclonal Antibody
Anti-IPAF Rabbit Polyclonal Antibody

100 µg

Price
494,00 €
Log in for availability
1 * 100 µG
Product Details & Documents

Ipaf Antibody: Apoptosis is related to many diseases and induced by a family of cell death receptors and their ligands. Cell death signals are transduced by death domain containing adaptor molecules and proteases including several members of the caspase family. Another family of proteins that functions as a critical regulator of apoptosis and NF kappa signaling pathways is the CED-4/Apaf-1 (apoptosis protein activating factor-1) protein family. Ipaf (ICE protease activating factor) is a CED-4/Apaf-1 family member that activates caspase-1/ICE and can induce apoptosis in human cells in a caspase-1 dependent manner. Ipaf and caspase-1 are thought to interact with each other through the association of the Ipaf amino-terminal CARD (caspase recruitment domain) and amino-terminal CARD of caspase-1.

Type:
Antigen: IPAF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Documents

Specifications
  • Size:
    100 µg
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Purification:
    Affinity chromatography purified via peptide column
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Host:
    Rabbit
  • Cat. No.:
    PRSI3107
  • Reactivity:
    Human
  • Immunogen:
    Ipaf antibody was raised against a synthetic peptide corresponding to amino acids near the C-terminus of human Ipaf.
  • ImmunoChemistry:
    Yes
  • Conjugation:
    Unconjugated
  • Tested applications:
    E, WB, ICC, IF
  • Antigen symbol:
    IPAF
  • Storage buffer:
    PBS containing 0.02% sodium azide
  • ELISA:
    Yes