Order Entry
Italy
ContactUsLinkComponent
Anti-PMAIP1 Rabbit Polyclonal Antibody
Anti-PMAIP1 Rabbit Polyclonal Antibody
Catalog # PRSI2437
Supplier:  ProSci Inc.
undefined
Anti-PMAIP1 Rabbit Polyclonal Antibody
Catalog # PRSI2437
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Phorbol-12-myristate-13-acetate-induced protein 1
  • Antigen symbol:
    PMAIP1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Antigen synonyms:
    Immediate-early-response protein APR|PMA-induced protein 1|NOXA|Protein Noxa|Phorbol-12-myristate-13-acetate-induced protein 1
  • Storage buffer:
    PBS containing 0.02% sodium azide
  • Immunogen:
    Noxa antibody was raised against a synthetic peptide corresponding to 17 amino acids at the amino terminus of mouse Noxa.
  • Tested applications:
    E, WB, IHC, IF
  • Purification:
    Immunoaffinity chromatography purified IgG
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Noxa Antibody: Apoptosis is related to many diseases and development. The p53 tumor-suppressor protein induces apoptosis through transcriptional activation of several genes including p53R2, p53AIP1, and PUMA. A new p53 target gene, Noxa, was recently identified, which encodes a protein belonging to the subfamily of BH3-only proapoptic proteins. Noxa and PUMA are both transcriptional targets of p53 and BH3-only proteins. X-ray irradiation increased p53-dependent Noxa mRNA and protein levels. Noxa, when ectopically expressed, interacted with anti-apoptotic Bcl-2 family members, resulting in the activation of caspase-9. Noxa, like PUMA, localized to mitochondria and induces apoptosis in response to p53. Noxa and PUMA may represent direct mediators of p53-induced apoptosis. Increased levels of p53 and its target gene Noxa was found in the impaired tumor development.

Type:
Antigen: PMAIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat