
Imidazolidinyl urea ≥98% (by titrimetric analysis) (N 26-28%)
:39236-46-9
:
:C₁₁H₁₆N₈O₈
:MFCD00221482
:388.30 g/mol
:141…143 °C
:254-372-6
:GHS07
- Appearance (Color):White to Pale Yellow
- Form:Solid
- Assay (Titration):98%
- Elemental Analysis:28%≥N≥26%
- Loss on Drying:≤5.0%
Frequently Bought Together