Order Entry
Italy
ContactUsLinkComponent
Single element standards for oil analysis, barium, Conostan®
Single element standards for oil analysis, barium, Conostan®
Catalog # PROOCON150-500-565
CAS Number:  
Single element standards for oil analysis, barium, Conostan®
Catalog # PROOCON150-500-565
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    Base Oil 20
  • Pk:
    50 g
  • Element:
    Barium
  • Symbol:
    Ba

Specifications

About this item