Order Entry
Italy
ContactUsLinkComponent
Organic reference standards, 10-hydroxyamitriptyline, LoGiCal®
Organic reference standards, 10-hydroxyamitriptyline, LoGiCal®
Catalog # PROOLGCFOR0062.05
CAS Number:  
Organic reference standards, 10-hydroxyamitriptyline, LoGiCal®
Catalog # PROOLGCFOR0062.05
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Reference standard
  • CAS No.:
    1159-82-6
  • Description:
    10-Hydroxyamitriptyline
  • Formula:
    C₂₀H₂₃NO
  • Description:
    10-Hydroxyamitriptyline

Specifications

About this item