Order Entry
Italy
ContactUsLinkComponent
 
Trazodone Related Compounds, Reference Standard
 :  USP
 
Trazodone Related Compounds, Reference Standard
 :  USP

 

  • CAS No.:
    25332-39-2
  • Formula:
    C₁₉H₂₃Cl₂N₅O
  • Description:
    Trazodone Hydrochloride
  • Synonym(s):
    2-{3-[4-(3-bromophenyl)piperazin-1-yl]propyl}-[1,2,4]triazolo[4,3-a]pyridin-3(2H)-one hydrochloride

 

Frequently Bought Together