Order Entry
Italy
ContactUsLinkComponent
Lockout Bags
Lockout Bags
Catalog # BRDY065780
Supplier:  Brady
undefined
Lockout Bags
Catalog # BRDY065780
Supplier:  Brady
Specifications
  • Description:
    Lockout bag, yellow
  • Dimensions:
    248×457 mm
  • Pk:
    1
Specifications
About this item
Lockout bag made of rugged canvas with drawstrings.
  • Can be locked closed Legend: LOCK OUT FOR SAFETY
For securing plugs on portable applications, cables on pneumatic connectors, crane pendants.