Ropivacaine, Reference Standards
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
- Catalog No:
- 1605497.
- 1605500.
- 1605512.
- 1605523.
- Description:
- Ropivacaine
- Ropivacaine Hydrochloride
- Ropivacaine Related Compound A
- Ropivacaine Related Compound B
- Synonym(s):
- 2,6-dimethylaniline hydrochloride
- (R)-N-(2,6-Dimethylphenyl)-1-propylpiperidine-2-carboxamide hydrochloride monohydrate
- CAS No.:
- 84057-95-4
- 132112-35-7
- 21436-98-6
- 112773-90-7
- Formula:
- C₁₇H₂₆N₂O
- C₁₇H₂₉ClN₂O₂
- C₈H₁₂ClN
- C₁₇H₂₇ClN₂O
Product Family Options
- DescriptionStandardsPack typePk
- RopivacaineUSP reference standardVial200 mg1 * 200 mg396,00 €
Specifications
Cat. No.1605497.DescriptionRopivacaineSynonym(s)CAS No.84057-95-4FormulaC₁₇H₂₆N₂O - Ropivacaine HydrochlorideUSP reference standardVial200 mg1 * 200 mg406,00 €
Specifications
Cat. No.1605500.DescriptionRopivacaine HydrochlorideSynonym(s)CAS No.132112-35-7FormulaC₁₇H₂₉ClN₂O₂ - Ropivacaine Related Compound AUSP reference standardVial25 mg1 * 25 mg1 150,00 €
Specifications
Cat. No.1605512.DescriptionRopivacaine Related Compound ASynonym(s)2,6-dimethylaniline hydrochlorideCAS No.21436-98-6FormulaC₈H₁₂ClN - Ropivacaine Related Compound BUSP reference standardVial50 mg1 * 50 mg1 360,00 €
Specifications
Cat. No.1605523.DescriptionRopivacaine Related Compound BSynonym(s)(R)-N-(2,6-Dimethylphenyl)-1-propylpiperidine-2-carboxamide hydrochloride monohydrateCAS No.112773-90-7FormulaC₁₇H₂₇ClN₂O



