Anti-Gc Rabbit Polyclonal Antibody
Catalog # BSBTA03364-2
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Gc
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Rat,Mouse
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Gc detection. Tested with WB, Direct ELISA in Mouse;Rat.
Type: Primary
Antigen: Gc
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse;Rat