Anti-Frizzled 4 Rabbit Polyclonal Antibody
BSBTA02191-1
- Antibody type:Primary
- Antigen name:Frizzled 4
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for Frizzled 4 detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Type: Primary
Antigen: Frizzled 4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat