Anti-TFPI2 Rabbit Polyclonal Antibody
BSBTA03697-1
- Antibody type:Primary
- Antigen name:Tissue factor pathway inhibitor 2
- Antigen symbol:TFPI2
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for Tissue factor pathway inhibitor 2(TFPI2) detection. Tested with WB, IHC-P in Human;Mouse.
Type: Primary
Antigen: TFPI2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse