Anti-ATF6 Rabbit Polyclonal Antibody
Catalog # BSBTA00655
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Cyclic AMP-dependent transcription factor ATF-6 alpha
- Antigen symbol:ATF6
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Cyclic AMP-dependent transcription factor ATF-6 alpha(ATF6) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Type: Primary
Antigen: ATF6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat