About this item
pET-41 series is designed for cloning and high-level expression of peptide sequences fused with the 220 aa GST•Tag™ protein. The cloning/expression region of the coding strand transcribed by T7 RNA polymerase. The f1 origin is oriented so that infection with helper phage will produce virions containing single stranded DNA that corresponds to the coding strand.