Order Entry
Italy
Orders LinkContactUsLinkComponent
Oxytocin, Reference Standards

Oxytocin, Reference Standards

 :   USP
Oxytocin, Reference Standards
 :  USP
 
Oxytocin, Reference Standards

 

 

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Caution: Cold Shipment required

 

 

 

  • Catalog No:
  • 1491300.
  • 1491296.
  • Description:
  • Oxytocin
  • Oxytocin Identication
  • CAS No.:
  • 50-56-6
  • 6233-83-6
  • Formula:
  • C₄₃H₆₆N₁₂O₁₂S₂
  • C₄₅H₇₀N₁₂O₁₄S₂
  • UN No.:
  • 2811
  • 2811

 

Product Family Options

 

  • Description
    Standards
    Pk Info
    Pack type
    Pk
     
     
  • Oxytocin
    USP reference standard
    1.81 mg
    Vial
    1 Vial
       
       
    1 * 1 Vial
    1 * 1 Vial - 550,00 €
    550,00 €
    1 * 1 Vial
       550,00 €

     
     
    Specifications
    Cat. No.
    1491300.
    Description
    Oxytocin
    CAS No.
    50-56-6
    Formula
    C₄₃H₆₆N₁₂O₁₂S₂
    UN No.
    2811
  • Oxytocin Identication
    USP reference standard
    Vial
    15 mg
       
       
    1 * 15 mg
    1 * 15 mg - 648,00 €
    648,00 €
    1 * 15 mg
       648,00 €

     
     
    Specifications
    Cat. No.
    1491296.
    Description
    Oxytocin Identication
    CAS No.
    6233-83-6
    Formula
    C₄₅H₇₀N₁₂O₁₄S₂
    UN No.
    2811
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.