Valacyclovir Related Compounds, Reference Standard
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
6 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1707855.
- 1707861.
- 1707872.
- 1707883.
- 1707894.
- 1707828.
- Description:
- Valacyclovir Related Compound C
- Valacyclovir Related Compound D
- Valacyclovir Related Compound E
- Valacyclovir Related Compound F
- Valacyclovir Related Compound G
- Valacyclovir Related Compound M
- Synonym(s):
- 2-[(2-amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]ethyl N-methyl-L-valinate hydrochloride
- 2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9yl)methoxy]ethyl N-ethyl-L-valinate
- 2-[(2-amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]ethyl N-[(benzyloxy)carbonyl]-L-valinate
- 2-hydroxyethyl valinate para-toluenesulfonate salt
- N,N-dimethylpyridin-4-amine
- 9-[(2-Hydroxyethoxy)methyl]guanine N-formyl-l-valinate
- CAS No.:
- 1346617-39-7
- 1346747-69-0
- 124832-31-1
- 86150-61-0
- 1122-58-3
- 847670-62-6
- Formula:
- C₁₄H₂₃ClN₆O₄
- C₁₅H₂₄N₆O₄
- C₂₁H₂₆N₆O₆
- C₁₄H₂₃NO₆S
- C₇H₁₀N₂
- UN No.:
- 2928
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1707855.Valacyclovir Related Compound CValacyclovir Related Compound CUSP reference standardVial10 mgLog in for availability1 * 10 mg1 360,00 €Specifications:
More
Specifications
Cat. No.1707855.DescriptionValacyclovir Related Compound CSynonym(s)2-[(2-amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]ethyl N-methyl-L-valinate hydrochlorideCAS No.1346617-39-7FormulaC₁₄H₂₃ClN₆O₄UN No. - 1707861.Valacyclovir Related Compound DValacyclovir Related Compound DUSP reference standardVial15 mgLog in for availability1 * 15 mg1 210,00 €Specifications:
More
Specifications
Cat. No.1707861.DescriptionValacyclovir Related Compound DSynonym(s)2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9yl)methoxy]ethyl N-ethyl-L-valinateCAS No.1346747-69-0FormulaC₁₅H₂₄N₆O₄UN No. - 1707872.Valacyclovir Related Compound EValacyclovir Related Compound EUSP reference standardVial25 mgLog in for availability1 * 25 mg1 240,00 €Specifications:
More
Specifications
Cat. No.1707872.DescriptionValacyclovir Related Compound ESynonym(s)2-[(2-amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]ethyl N-[(benzyloxy)carbonyl]-L-valinateCAS No.124832-31-1FormulaC₂₁H₂₆N₆O₆UN No. - 1707883.Valacyclovir Related Compound FValacyclovir Related Compound FUSP reference standardVial25 mgLog in for availability1 * 25 mg1 240,00 €Specifications:
More
Specifications
Cat. No.1707883.DescriptionValacyclovir Related Compound FSynonym(s)2-hydroxyethyl valinate para-toluenesulfonate saltCAS No.86150-61-0FormulaC₁₄H₂₃NO₆SUN No. - 1707894.Valacyclovir Related Compound GValacyclovir Related Compound GUSP reference standardVial50 mgLog in for availability1 * 50 mg1 260,00 €Specifications:
More
Specifications
Cat. No.1707894.DescriptionValacyclovir Related Compound GSynonym(s)N,N-dimethylpyridin-4-amineCAS No.1122-58-3FormulaC₇H₁₀N₂UN No.2928 - 1707828.Valacyclovir Related Compound MValacyclovir Related Compound MUSP reference standardVial15 mgLog in for availability1 * 15 mg1 140,00 €Specifications:
More
Specifications
Cat. No.1707828.DescriptionValacyclovir Related Compound MSynonym(s)9-[(2-Hydroxyethoxy)methyl]guanine N-formyl-l-valinateCAS No.847670-62-6FormulaUN No.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



