Order Entry
Italy
Orders LinkContactUsLinkComponent
Amikacin, Reference Standards

Amikacin, Reference Standards

Supplier:   USP
Amikacin, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1019508.
  • 1019494.
  • Description:
  • Amikacin
  • Amikacin Sulfate
  • CAS No.:
  • 37517-28-5
  • 39831-55-5
  • Formula:
  • C₂₂H₄₃N₅O₁₃
  • C₂₂H₄₇N₅O₂₁S₂

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1019508.
    Amikacin
    Amikacin
    USP reference standard
    Vial
    300 mg
    Log in for availability
    Log in for availability
    1 * 300 mg
    1 * 300 mg - 418,00 €
    418,00 €
    1 * 300 mg
       418,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1019508.
    Description
    Amikacin
    CAS No.
    37517-28-5
    Formula
    C₂₂H₄₃N₅O₁₃
  • 1019494.
    Amikacin Sulfate
    Amikacin Sulfate
    USP reference standard
    Vial
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 376,00 €
    376,00 €
    1 * 100 mg
       376,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1019494.
    Description
    Amikacin Sulfate
    CAS No.
    39831-55-5
    Formula
    C₂₂H₄₇N₅O₂₁S₂