Order Entry
Italy
Orders LinkContactUsLinkComponent
Norelgestromin, Reference Standards

Norelgestromin, Reference Standards

Supplier:   USP
Norelgestromin, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1468454.
  • 1468465.
  • Description:
  • Norelgestromin
  • Norelgestromin Related Compound A
  • Synonym(s):
  • 13-Ethyl-17-hydroxy-18,19-dinor-17α-pregn-5(6)-en-20-yn-3-one oxime
  • CAS No.:
  • 53016-31-2
  • Formula:
  • C₂₁H₂₉NO₂

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1468454.
    Norelgestromin
    Norelgestromin
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 646,00 €
    646,00 €
    1 * 200 mg
       646,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1468454.
    Description
    Norelgestromin
    Synonym(s)
    CAS No.
    53016-31-2
    Formula
    C₂₁H₂₉NO₂
  • 1468465.
    Norelgestromin Related Compound A
    Norelgestromin Related Compound A
    USP reference standard
    Vial
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - 1 330,00 €
    1 330,00 €
    1 * 15 mg
       1 330,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1468465.
    Description
    Norelgestromin Related Compound A
    Synonym(s)
    13-Ethyl-17-hydroxy-18,19-dinor-17α-pregn-5(6)-en-20-yn-3-one oxime
    CAS No.
    Formula