mono-Fmoc ethylene diamine hydrochloride ≥98% (by TLC), Novabiochem®
:391624-46-7
:166410-32-8
:C₁₇H₁₈N₂O₂·HCl
:MFCD00830741
:318.80 g/mol
- Colour:White to slight yellow to beige
- Appearance of substance:Powder
- Identity (IR):Passes test
- Purity (TLC(CMA2)):≥ 98%
- Storage:Store at 2...8 °C
Frequently Bought Together