Order Entry
Italy
ContactUsLinkComponent
Organic reference standards, tenoxicam, Ph. Eur., RS
Organic reference standards, tenoxicam, Ph. Eur., RS
Supplier:  EDQM
Certificates
Organic reference standards, tenoxicam, Ph. Eur., RS
Supplier:  EDQM

Specifications

  • CAS No.:
    59804-37-4
  • Formula:
    C₁₃H₁₁N₃O₄S₂
  • Description:
    Tenoxicam

Specifications

Frequently Bought Together