Order Entry
Italy
ContactUsLinkComponent
Organic reference standard, salmeterols, Ph. Eur., RS
Organic reference standard, salmeterols, Ph. Eur., RS
Supplier:  EDQM
Certificates
Organic reference standard, salmeterols, Ph. Eur., RS
Supplier:  EDQM

Specifications

  • CAS No.:
    94749-08-3
  • Formula:
    C₃₆H₄₅NO₇
  • Description:
    Salmeterol xinafoate for system suitability

Specifications

Frequently Bought Together