Organic reference standards, medroxyprogesterone acetate, Ph. Eur. RS
Supplier: EDQM
Certificates
About this item
Specifications
- CAS No.:71-58-9
- Formula:C₂₄H₃₄O₄
- Description:Medroxyprogesterone acetate
Specifications
Frequently Bought Together