Order Entry
Italy
ContactUsLinkComponent
Organic reference standards, orphenadrine, Ph. Eur. RS
Organic reference standards, orphenadrine, Ph. Eur. RS
Supplier:  EDQM
Certificates
Organic reference standards, orphenadrine, Ph. Eur. RS
Supplier:  EDQM

Specifications

  • CAS No.:
    21945-86-8
  • Formula:
    C₁₈H₂₃NO
  • Description:
    Orphenadrine impurity E, (RS)-N,N-dimethyl-2-[(3-methylphenyl)phenylmethoxy] ethanamine (metamethylbenzyl isomer)

Specifications

Frequently Bought Together