Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Inserisci ordine
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodio dodecilbenzenesulfonato
Sodio dodecilbenzenesulfonato 65% in water, soft type
Sodio dodecilbenzenesulfonato 65% in water, soft type
Sodio dodecilbenzenesulfonato 65% in water, soft type
ICNA0520331125
:
MP Biomedicals
:
Sodio dodecilbenzenesulfonato 65% in water, soft type
ICNA0520331125
:
MP Biomedicals
:
Conf:
25 g
Seq:
3308
Form:
White creamy paste
Detection method:
FTIR
:
:
348.48 g/mol
:
3077
:
,
:
GHS05
:
GHS07
:
25155-30-0
:
C₁₈H₂₉NaO₃S
:
MFCD00011508