Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Bromate-Bromide
Bromate-Bromide 0.05 M Br2 (0.1 N) volumetric solution, Fluka™
Bromide bromate 0.05 M Br2 (0.1 N) volumetric solution, Fluka™
Bromide bromate 0.05 M Br2 (0.1 N) volumetric solution, Fluka™
35006-1L
:
Honeywell Chemicals
:
Bromide bromate 0.05 M Br2 (0.1 N) volumetric solution, Fluka™
35006-1L
:
Honeywell Chemicals
:
Pack type:
Glass bottle
Pk:
1 L
Seq:
5037
Pk Info:
Glass bottle
Factor:
0.998 - 1.002
:
:
1.01 g/cm³
:
GHS08
:
Ambient
:
7758-01-2
:
KBrO₃/KBr