Order Entry
Italy
ContactUsLinkComponent
Anti-CD45 Rat Monoclonal Antibody (Biotin) [clone: 30-F11]
Anti-CD45 Rat Monoclonal Antibody (Biotin) [clone: 30-F11]
Supplier:  Tonbo Biosciences
Certificates
Anti-CD45 Rat Monoclonal Antibody (Biotin) [clone: 30-F11]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 30-0451-U100
  • 30-0451-U025
  • Antigen symbol:
  • CD45
  • CD45
  • Antigen synonyms:
  • Leukocyte Common Antigen, LCA, Ly-5
  • Leukocyte Common Antigen, LCA, Ly-5
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • 30-F11
  • 30-F11
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 19264
  • 19264
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • 2-8°C
  • 2-8°C

Specifications

Frequently Bought Together