Anti-IgG Donkey Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 72-8097-M001
Supplier: Tonbo Biosciences
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Donkey
- ImmunoChemistry:Yes
- Reactivity:Mouse
- Western blot:Yes
- Epitope:Heavy and light chain
- Storage temperature:Store freeze-dried powder at 2-8 °C. Once rehydrated and in solution store at 2-8 °C.
- Pk:1 mg
Specifications
About this item
Donkey anti-Mouse IgG (H&L) - Affinity Pure, HRP Conjugate
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and light chain
Host: Donkey
Isotype:
Reactivity: Mouse