Chemical resistant gloves, Ultranitril 491
Supplier: MAPA
Certificates
About this item
These anatomically designed gloves with flock lining provide good mechanical resistance and long-lasting chemical protection.
- Dexterity and comfort thanks to the anatomical shape and the quality of the flock lining
- Longer service life: Excellent mechanical resistance (abrasion, perforation)
- Embossed texture for improved grip
Typical applications include: Automotive/mechanical industry, treating/degreasing metals with solvents, machining parts using cutting oil, maintenance, timber treatment and finishing, application of pesticides, cleaning of printing-press rollers, chemical industry, manufacture and application of paints and varnishes.
Certifications: EN 388:2016 (3101X), EN ISO 374-1 Type A (AJKOPT), EN ISO 374-5 VIRUS; ISO 18889 G2; Cat. III
Specifications
- Catalog No:
- MAPA34491260
- MAPA34491266
- MAPA34491268
- MAPA34491267
- MAPA34491269
- Model:
- Ultranitril 491
- Ultranitril 491
- Ultranitril 491
- Ultranitril 491
- Ultranitril 491
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Cotton
- Cotton
- Cotton
- Cotton
- Cotton
- Finish:
- Embossed
- Embossed
- Embossed
- Embossed
- Embossed
- Cuff style:
- Turned & lined
- Turned & lined
- Turned & lined
- Turned & lined
- Turned & lined
- EN norm:
- EN 388:2016 (3101X), EN ISO 374-1 Type A (AJKOPT), EN ISO 374-5 VIRUS
- EN 388:2016 (3101X), EN ISO 374-1 Type A (AJKOPT), EN ISO 374-5 VIRUS
- EN 388:2016 (3101X), EN ISO 374-1 Type A (AJKOPT), EN ISO 374-5 VIRUS
- EN 388:2016 (3101X), EN ISO 374-1 Type A (AJKOPT), EN ISO 374-5 VIRUS
- EN 388:2016 (3101X), EN ISO 374-1 Type A (AJKOPT), EN ISO 374-5 VIRUS