Industrial gloves, Ultrane 551
Supplier: MAPA
Certificates
About this item
These seamless knitted gloves with PU foam coating on palm and fingertips provide excellent abrasion resistance.
- Excellent tactile sensitivity due to reduced glove thickness
- Absorption of perspiration ensured by the cellular structure of the PU coating
- Ventilated back for additional wearing comfort
Typical applications include: Automotive/mechanical industry, sorting small parts, fitting small screws and fasteners, electronic component assembly (displays, LEDs, etc.), high-frequency antennae riveting, microprocessor handling, food industry, cosmetics and pharmaceutical industry, construction industry (metalworkers/locksmiths), installing and repairing locks, maintaining pipes, heating equipment, laying wood, panelling or moudling.
Certifications: EN 388:2016 (4131X); Cat. II
Specifications
- Catalog No:
- MAPA34551039
- MAPA34551038
- MAPA34551037
- MAPA34551031
- MAPA34551036
- MAPA34551030
- Model:
- Ultrane 551
- Ultrane 551
- Ultrane 551
- Ultrane 551
- Ultrane 551
- Ultrane 551
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Finish:
- 3/4 coating
- 3/4 coating
- 3/4 coating
- 3/4 coating
- 3/4 coating
- 3/4 coating
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4131X)
- EN 388:2016 (4131X)
- EN 388:2016 (4131X)
- EN 388:2016 (4131X)
- EN 388:2016 (4131X)
- EN 388:2016 (4131X)