L(+)-Tartaric acid Ph. Eur., Macron Fine Chemicals™
Catalog # P107-33
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
Merck Index:13,09158
Boiling Pt:399 °C (1013 hPa)
Melting Pt:168…170 °C
Density:1.759 g/cm³ (20 °C)
Flash Pt:150 °C (closed cup)
:150.09 g/mol
Storage Temperature:Ambient
EINECS:201-766-0
REACH:01-2119537204-47
:GHS05
CAS Number:87-69-4
Formula:C₄H₆O₆
MDL Number:MFCD00064207