Order Entry
Italy
ContactUsLinkComponent
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate free acid ≥98%, purified, J.T.Baker®
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate free acid ≥98%, purified, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate free acid ≥98%, purified, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    ≥98%
  • Assay (C₆H₁₃NO₄S·H₂O):
    ≥98 %
  • Appearance:
    Passes Test
  • Identification (by IR):
    Passes Test
  • RNase Activity:
    Passes Test
  • DNase Activity (None Detected):
    Passes Test
  • Protease Activity:
    Passes Test
  • Color of a 1M Alkaline Solution (Colorless):
    Passes Test
  • Loss on Drying at 130°C:
    7 - 10 %
  • Heavy Metals (as Pb):
    ≤2 ppm

Specification Test Results

Related Information

Frequently Bought Together